Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082047.1 | NP_001075516.1 | 1500 | 118 | ||
GenBank mRNA | AF109472.1 | - | 1500 | 118 |
Gene Symbol: LOC103344941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008253072.1 | XP_008251294.1 | 346 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus |