Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082048.1 | NP_001075517.1 | 8 - 9 | 961 | 118 | |
GenBank mRNA | AF118146.1 | - | 8 - 9 | 961 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Oryctolagus cuniculus Probe spans exons |