Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082065.1 | NP_001075534.1 | 17 - 18 | 1778 | 62 | |
GenBank mRNA | AF170528.1 | - | 17 - 18 | 1778 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |