Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082066.1 | NP_001075535.1 | 12 - 13 | 2016 | 93 | |
XM_008271093.2 | XP_008269315.1 | 12 - 13 | 2803 | 93 | ||
GenBank mRNA | AF170529.1 | - | 12 - 13 | 2016 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |