Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR1I2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082067.2 | NP_001075536.1 | 6 - 7 | 1125 | 59 | |
XM_008266819.2 | XP_008265041.1 | 6 - 7 | 1310 | 59 | ||
GenBank mRNA | AF182217.1 | - | 6 - 7 | 1126 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |