Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082070.1 | NP_001075539.1 | 20 - 21 | 2401 | 78 | |
GenBank mRNA | AB009312.1 | - | 19 - 20 | 78 | ||
AF201702.1 | - | 20 - 21 | 2401 | 78 | ||
AF321818.1 | - | 19 - 20 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |