Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082075.1 | NP_001075544.1 | 4 - 5 | 318 | 71 | |
XM_008254831.2 | XP_008253053.1 | 5 - 6 | 379 | 71 | ||
GenBank mRNA | AF209725.1 | - | 4 - 5 | 318 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons |