Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082088.1 | NP_001075557.1 | 1 - 2 | 49 | 72 | |
XM_008263947.2 | XP_008262169.1 | 1 - 2 | 123 | 72 | ||
GenBank mRNA | AF276980.1 | - | 1 - 2 | 49 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |