Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082103.1 | NP_001075572.1 | 7 - 8 | 895 | 121 | |
GenBank mRNA | AF176914.5 | - | 7 - 8 | 895 | 121 | |
AF332576.4 | - | 7 - 8 | 895 | 121 | ||
M29852.1 | - | 7 - 8 | 886 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |