Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082118.1 | NP_001075587.1 | 3 - 4 | 1375 | 87 | |
GenBank mRNA | AF493547.1 | - | 3 - 4 | 1335 | 87 | |
AF508735.1 | - | 3 - 4 | 1375 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |