Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082127.1 | NP_001075596.1 | 628 | 61 | ||
GenBank mRNA | AJ242871.1 | - | 628 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus |