Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082153.1 | NP_001075622.1 | 28 - 29 | 4346 | 77 | |
XM_008263960.2 | XP_008262182.1 | 28 - 29 | 4700 | 77 | ||
GenBank mRNA | AY212921.1 | - | 28 - 29 | 4346 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |