Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082181.1 | NP_001075650.1 | 5 - 6 | 613 | 150 | |
GenBank mRNA | AY864853.1 | - | 5 - 6 | 613 | 150 | |
DQ217840.1 | - | 3 - 4 | 312 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |