Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082190.1 | NP_001075659.1 | 4 - 5 | 524 | 89 | |
GenBank mRNA | D13668.1 | - | 4 - 5 | 524 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |