Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082214.1 | NP_001075683.1 | 2 - 3 | 264 | 64 | |
XM_008263962.2 | XP_008262184.1 | 2 - 3 | 320 | 64 | ||
GenBank mRNA | DQ120011.1 | - | 2 - 3 | 264 | 64 | |
GQ848313.1 | - | 2 - 3 | 264 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Oryctolagus cuniculus Probe spans exons |