Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082225.1 | NP_001075694.1 | 2 - 3 | 583 | 59 | |
GenBank mRNA | AB001934.1 | - | 2 - 3 | 179 | 59 | |
AB009313.1 | - | 2 - 3 | 222 | 59 | ||
AF107300.1 | - | 2 - 3 | 208 | 59 | ||
AY829265.1 | - | 2 - 3 | 200 | 59 | ||
DQ821756.2 | - | 2 - 3 | 583 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |