Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082255.1 | NP_001075724.1 | 2 - 3 | 189 | 62 | |
XM_008263970.2 | XP_008262192.1 | 3 - 4 | 353 | 62 | ||
XM_017345487.1 | XP_017200976.1 | 3 - 4 | 353 | 62 | ||
GenBank mRNA | L26981.1 | - | 2 - 3 | 189 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |