Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082284.1 | NP_001075753.1 | 5 - 6 | 708 | 72 | |
XM_008263973.2 | XP_008262195.2 | 5 - 6 | 752 | 72 | ||
GenBank mRNA | M32029.1 | - | 5 - 6 | 708 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |