Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PON1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082297.1 | NP_001075766.1 | 2 - 3 | 197 | 84 | |
GenBank mRNA | AF220941.1 | - | 1 - 2 | 71 | 84 | |
AF220942.1 | - | 2 - 3 | 168 | 84 | ||
AF220943.1 | - | 2 - 3 | 168 | 84 | ||
M63011.1 | - | 2 - 3 | 197 | 84 | ||
S64616.1 | - | 2 - 3 | 197 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |