Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082303.1 | NP_001075772.1 | 1 - 2 | 799 | 147 | |
GenBank mRNA | M65109.1 | - | 1 - 2 | 799 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |