Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082310.1 | NP_001075779.1 | 6 - 7 | 765 | 87 | |
GenBank mRNA | M84474.1 | - | 6 - 7 | 765 | 87 | |
M84988.1 | - | 6 - 7 | 765 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |