Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082322.1 | NP_001075791.1 | 601 | 60 | ||
GenBank mRNA | S51597.1 | - | 601 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Oryctolagus cuniculus |