Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082331.1 | NP_001075800.1 | 3 - 4 | 1152 | 69 | |
GenBank mRNA | S82448.1 | - | 3 - 4 | 1152 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Oryctolagus cuniculus Probe spans exons |