Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Oc03398128_m1
See other SLMAP GE Assays ›
Gene Symbol:
SLMAP
Gene Name:
sarcolemma associated protein
Gene Aliases:
SLAP
Chromosome Location:
Chr.9: 24582901 - 24751719 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID
Oc03398128_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SLMAP
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001082348.2
NP_001075817.2
5 - 6
519
115
XM_008260454.2
XP_008258676.1
6 - 7
1755
115
XM_008260455.2
XP_008258677.1
6 - 7
1749
115
XM_008260456.2
XP_008258678.1
6 - 7
1755
115
XM_008260457.2
XP_008258679.1
6 - 7
1755
115
XM_008260458.2
XP_008258680.1
6 - 7
1755
115
XM_008260459.2
XP_008258681.1
6 - 7
1749
115
XM_008260460.2
XP_008258682.1
6 - 7
1755
115
XM_008260461.2
XP_008258683.1
6 - 7
1749
115
XM_008260462.2
XP_008258684.1
6 - 7
1749
115
XM_008260463.2
XP_008258685.1
6 - 7
1755
115
XM_008260464.2
XP_008258686.1
6 - 7
1755
115
XM_008260465.2
XP_008258687.1
6 - 7
1755
115
XM_008260466.2
XP_008258688.1
6 - 7
1757
115
XM_008260467.2
XP_008258689.1
6 - 7
1754
115
XM_008260468.2
XP_008258690.1
6 - 7
1748
115
XM_008260470.2
XP_008258692.1
6 - 7
1757
115
XM_008260471.2
XP_008258693.1
6 - 7
1750
115
XM_008260472.2
XP_008258694.1
6 - 7
1756
115
XM_008260473.2
XP_008258695.1
6 - 7
1750
115
XM_017343638.1
XP_017199127.1
6 - 7
1749
115
XM_017343639.1
XP_017199128.1
6 - 7
1760
115
GenBank mRNA
U21157.1
-
5 - 6
880
115
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Oryctolagus cuniculus
Probe spans exons
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0