Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHBG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082370.1 | NP_001075839.1 | 6 - 7 | 892 | 65 | |
GenBank mRNA | U57553.1 | - | 6 - 7 | 858 | 65 | |
X12661.1 | - | 534 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |