Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082393.1 | NP_001075862.1 | 1 - 2 | 368 | 95 | |
XM_008271101.2 | XP_008269323.1 | 2 - 3 | 534 | 95 | ||
GenBank mRNA | M32231.1 | - | 1 - 2 | 275 | 95 | |
X56031.1 | - | 368 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |