Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082393.1 | NP_001075862.1 | 2 - 3 | 444 | 57 | |
XM_008271101.2 | XP_008269323.1 | 3 - 4 | 610 | 57 | ||
GenBank mRNA | M32231.1 | - | 2 - 3 | 351 | 57 | |
X56031.1 | - | 444 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |