Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082700.1 | NP_001076169.1 | 14 - 15 | 1219 | 90 | |
XM_017346648.1 | XP_017202137.1 | 13 - 14 | 1186 | 90 | ||
GenBank mRNA | U32507.1 | - | 14 - 15 | 1219 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |