Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082700.1 | NP_001076169.1 | 16 - 17 | 1519 | 76 | |
XM_017346648.1 | XP_017202137.1 | 15 - 16 | 1486 | 76 | ||
GenBank mRNA | U32507.1 | - | 16 - 17 | 1519 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |