Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082700.1 | NP_001076169.1 | 17 - 18 | 1555 | 117 | |
XM_017346648.1 | XP_017202137.1 | 16 - 17 | 1522 | 117 | ||
GenBank mRNA | U32507.1 | - | 17 - 18 | 1555 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |