Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082712.1 | NP_001076181.1 | 3 - 4 | 225 | 63 | |
XM_008268542.2 | XP_008266764.1 | 3 - 4 | 286 | 63 | ||
XM_008268549.2 | XP_008266771.2 | 3 - 4 | 660 | 63 | ||
GenBank mRNA | AF040387.1 | - | 3 - 4 | 225 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Inventoried Probe spans exons |