Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082732.2 | NP_001076201.1 | 2178 | 140 | ||
XM_008273277.2 | XP_008271499.1 | 2066 | 140 | |||
GenBank mRNA | AY101394.1 | - | 2182 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Oryctolagus cuniculus |