Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082735.1 | NP_001076204.1 | 1158 | 73 | ||
GenBank mRNA | AY275542.1 | - | 1158 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Oryctolagus cuniculus |