Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082752.1 | NP_001076221.1 | 2 - 3 | 585 | 87 | |
GenBank mRNA | U23835.1 | - | 2 - 3 | 585 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |