Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RYR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082757.1 | NP_001076226.1 | 33 - 34 | 4910 | 83 | |
GenBank mRNA | M59743.1 | - | 33 - 34 | 4897 | 83 | |
U50465.1 | - | 33 - 34 | 4910 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |