Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082649.1 | NP_001076118.1 | 10 - 11 | 1434 | 113 | |
GenBank mRNA | AF028241.1 | - | 10 - 11 | 1434 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |