Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082671.1 | NP_001076140.1 | 1 - 2 | 1058 | 116 | |
NM_001122935.1 | NP_001116407.1 | 1 - 2 | 1078 | 116 | ||
XM_008263980.2 | XP_008262202.1 | 1 - 2 | 1108 | 116 | ||
XM_008263984.2 | XP_008262206.1 | 1 - 2 | 1107 | 116 | ||
XM_017345491.1 | XP_017200980.1 | 1 - 2 | 1107 | 116 | ||
XM_017345492.1 | XP_017200981.1 | 1 - 2 | 1107 | 116 | ||
GenBank mRNA | U04273.1 | - | 1 - 2 | 1058 | 116 | |
U04274.1 | - | 1 - 2 | 1078 | 116 | ||
U04276.1 | - | 1 - 2 | 365 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |