Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082707.1 | NP_001076176.1 | 1493 | 60 | ||
GenBank mRNA | AB022927.2 | - | 1493 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus |