Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082750.1 | NP_001076219.1 | 1 - 2 | 253 | 105 | |
XM_008259342.2 | XP_008257564.1 | 1 - 2 | 292 | 105 | ||
XM_017343246.1 | XP_017198735.1 | 1 - 2 | 205 | 105 | ||
GenBank mRNA | U09490.1 | - | 1 - 2 | 253 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |