Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082756.1 | NP_001076225.1 | 11 - 12 | 1131 | 77 | |
XM_017346650.1 | XP_017202139.1 | 11 - 12 | 1136 | 77 | ||
XM_017346651.1 | XP_017202140.1 | 9 - 10 | 872 | 77 | ||
GenBank mRNA | U46915.1 | - | 11 - 12 | 1131 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |