Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082759.2 | NP_001076228.2 | 6 - 7 | 233 | 92 | |
GenBank mRNA | X13043.1 | - | 233 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus Probe spans exons |