Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082772.1 | NP_001076241.1 | 7 - 8 | 720 | 60 | |
GenBank mRNA | X75451.1 | - | 720 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |