Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082772.1 | NP_001076241.1 | 9 - 10 | 905 | 93 | |
GenBank mRNA | X75451.1 | - | 905 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |