Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089311.1 | NP_001082780.1 | 1 - 2 | 62 | 99 | |
GenBank mRNA | AF276977.1 | - | 1 - 2 | 62 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |