Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089311.1 | NP_001082780.1 | 5 - 6 | 976 | 74 | |
GenBank mRNA | AF276977.1 | - | 5 - 6 | 976 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |