Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099964.2 | NP_001093434.1 | 5 - 6 | 1165 | 106 | |
GenBank mRNA | EF202603.1 | - | 5 - 6 | 1166 | 106 | |
EU982819.1 | - | 5 - 6 | 762 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Oryctolagus cuniculus Probe spans exons |