Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101689.1 | NP_001095159.1 | 1 - 2 | 681 | 99 | |
GenBank mRNA | X07995.1 | - | 1 - 2 | 681 | 99 |
Gene Symbol: LCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712428.3 | XP_002712474.1 | 1 - 2 | 639 | 99 | |
GenBank mRNA | Z27166.1 | - | 622 | 99 |
Gene Symbol: LOC100353264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712429.3 | XP_002712475.1 | 1 - 2 | 99 | ||
GenBank mRNA | Z27167.1 | - | 99 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |