Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101695.1 | NP_001095165.1 | 292 | 64 | ||
GenBank mRNA | X58727.1 | - | 292 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus |