Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101696.1 | NP_001095166.1 | 3 - 4 | 502 | 79 | |
GenBank mRNA | X59931.1 | - | 502 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |