Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101705.1 | NP_001095175.1 | 624 | 93 | ||
XM_008249640.2 | XP_008247862.1 | 787 | 93 | |||
GenBank mRNA | Y00684.1 | - | 624 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus |